Skip to Content
Merck
All Photos(1)

Key Documents

48160

Supelco

[2,2′]-Furildioxime monohydrate

for spectrophotometric det. of spectrophotometric determination of Au(II), Co(II), Cu(II), Ni(II), Pd(II), Re(VII), U(VI), ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8N2O4 · H2O
CAS Number:
Molecular Weight:
238.20
Beilstein:
85892
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

description

mixture of isomers (α-furildioxime:γ-furildioxime = ~80:20)

Assay

≥99.0% (TLC)
≥99.0%

quality

for spectrophotometric det. of spectrophotometric determination of Au(II), Co(II), Cu(II), Ni(II), Pd(II), Re(VII), U(VI)

technique(s)

UV/Vis spectroscopy: suitable

SMILES string

[H]O[H].O\N=C(c1ccco1)\C(=N\O)c2ccco2

Other Notes

Reagent for the spectrophotometric determination of Au(II), Co(II), Cu(II), Ni(II), Pd(II), Re(VII) and U(VI), a review

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R B Singh et al.
Talanta, 26(6), 425-444 (1979-06-01)
The uses of oximes as spectrophotometric reagents since 1953 are reviewed.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service