Skip to Content
Merck
All Photos(2)

Key Documents

37051

Supelco

Pyribencarb

PESTANAL®, analytical standard

Synonym(s):

Methyl {2-chloro-5-[(1E)-1-(6-methyl-2-pyridylmethoxyimino)ethyl]benzyl}carbamate

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
MXP 1,188.00

MXP 1,188.00


Estimated to ship onMay 21, 2025


Request a Bulk Order

Select a Size

Change View
25 MG
MXP 1,188.00

About This Item

Empirical Formula (Hill Notation):
C18H20ClN3O3
CAS Number:
Molecular Weight:
361.82
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

MXP 1,188.00


Estimated to ship onMay 21, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

COC(=O)NCc1cc(ccc1Cl)\C(C)=N\OCc2cccc(C)n2

InChI

1S/C18H20ClN3O3/c1-12-5-4-6-16(21-12)11-25-22-13(2)14-7-8-17(19)15(9-14)10-20-18(23)24-3/h4-9H,10-11H2,1-3H3,(H,20,23)/b22-13+

InChI key

CRFYLQMIDWBKRT-LPYMAVHISA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pyribencarb belongs to benzylcarbamate type of fungicides, which are active against a broad spectrum of economically important plant pathogenic fungi like Botrytis cinerea and Sclerotinia sclerotiorum.[1][2]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of a novel fungicide, pyribencarb
Takagati M, et al.
Journal of Pesticide Science, 39, 177-178 (2014)
Mechanism of action and selectivity of a novel fungicide, pyribencarb
Kataoka S, et al.
Journal of Pesticide Science, 35, 99-106 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service