Skip to Content
Merck
All Photos(2)

Key Documents

36717

Supelco

3,5-Dimethylphenol

PESTANAL®, analytical standard

Synonym(s):

5-Hydroxy-m-xylene, sym.-m-Xylenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C6H3OH
CAS Number:
Molecular Weight:
122.16
Beilstein:
774117
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

222 °C (lit.)

mp

61-64 °C (lit.)

application(s)

agriculture
cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

Cc1cc(C)cc(O)c1

InChI

1S/C8H10O/c1-6-3-7(2)5-8(9)4-6/h3-5,9H,1-2H3

InChI key

TUAMRELNJMMDMT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

179.6 °F

Flash Point(C)

82 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T C Sun et al.
Bone, 13(6), 443-446 (1992-01-01)
Substances that bind calcium are given to determine where and how fast bone is forming. Several vital dyes are used (tetracycline, calcein, alizarin, xylenol), but it is not known whether the histomorphometric results they provide are equivalent. This work tests
E C Cole et al.
Journal of applied microbiology, 95(4), 664-676 (2003-09-13)
To describe the relationship between antibiotic and antibacterial resistance in environmental and clinical bacteria from home environments across geographical locations, relative to the use or nonuse of antibacterial products, with a focus on target organisms recognized as potential human pathogens.
Oscar Núñez et al.
Journal of chromatography. A, 1175(1), 7-15 (2007-11-06)
The performance of a monolithic silica capillary column coated with poly(octadecyl methacrylate) (ODM column) for the reversed-phase liquid chromatographic separation of some polar and non-polar compounds was studied, and the results were compared to those obtained by using a monolithic
G Bieniek
Journal of chromatography. B, Biomedical applications, 682(1), 167-172 (1996-06-28)
An attempt was made to establish a method for the simultaneous determination of urinary concentrations of phenol, o-, p- and m-cresols, 1- and 2-naphthol and xylenol isomers by capillary gas chromatography. Urine samples were extracted after acid hydrolysis of glucuronides
S Laurent et al.
The quarterly journal of nuclear medicine and molecular imaging : official publication of the Italian Association of Nuclear Medicine (AIMN) [and] the International Association of Radiopharmacology (IAR), [and] Section of the Society of..., 53(6), 586-603 (2009-12-18)
Lanthanide complexes are more and more used in biomedical imaging as contrast agents (CA). The development of these paramagnetic complexes as CA for medical magnetic resonance imaging (MRI) and luminescent probes for optical imaging is very complementary. Gd complexes are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service