Skip to Content
Merck
All Photos(1)

Key Documents

364487

Sigma-Aldrich

N-(Diphenylmethylene)glycine tert-butyl ester

98%

Synonym(s):

tert-Butyl(diphenylmethyleneamino)acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2C=NCH2CO2C(CH3)3
CAS Number:
Molecular Weight:
295.38
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Quality Level

Assay

98%

form

powder, crystals or chunks

mp

114-115 °C (lit.)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

CC(C)(C)OC(=O)C\N=C(/c1ccccc1)c2ccccc2

InChI

1S/C19H21NO2/c1-19(2,3)22-17(21)14-20-18(15-10-6-4-7-11-15)16-12-8-5-9-13-16/h4-13H,14H2,1-3H3

InChI key

YSHDPXQDVKNPKA-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masakatsu Kanazawa et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 57(2), 303-308 (2015-11-14)
We recently developed a novel PET probe, 6-(11)C-methyl-m-tyrosine ((11)C-6MemTyr), for quantitative imaging of presynaptic dopamine synthesis in the living brain. In the present study, (11)C-6MemTyr was compared with β-(11)C-l-DOPA and 6-(18)F-fluoro-l-dopa ((18)F-FDOPA) in the brains of normal and Parkinson disease

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service