Skip to Content
Merck
All Photos(2)

Key Documents

32511

Supelco

Furazolidone-d4

VETRANAL®, analytical standard

Synonym(s):

3-(5-Nitrofurfurylideneamino)-2-oxazolidinone-d4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8D4H3N3O5
CAS Number:
Molecular Weight:
229.18
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

mass shift

M+4

SMILES string

[2H]C1([2H])OC(=O)N(\N=C\c2ccc(o2)[N+]([O-])=O)C1([2H])[2H]

InChI

1S/C8H7N3O5/c12-8-10(3-4-15-8)9-5-6-1-2-7(16-6)11(13)14/h1-2,5H,3-4H2/b9-5+/i3D2,4D2

InChI key

PLHJDBGFXBMTGZ-QIBFQDEWSA-N

General description

Furazolidone-d4 is an isotopically labeled compound typically used for the quantitation of furazolidone.

Application

Furazolidone-d4 may be used as an internal standard for the determination of furazolidone in pig muscle, liver and kidney tissues by high performance liquid chromatography-ultraviolet detection (HPLC-UV) and LC coupled to tandem mass spectrometry (MS/MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K M Cooper et al.
Food additives and contaminants, 22(5), 406-414 (2005-07-16)
Depletion of the nitrofuran antibiotics furazolidone, furaltadone, nitrofurantoin and nitrofurazone and their tissue-bound metabolites AOZ, AMOZ, AHD and SEM from pig muscle, liver and kidney tissues is described. Groups of pigs were given feed medicated with one of the nitrofuran

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service