Skip to Content
Merck
All Photos(1)

Key Documents

31091

Supelco

4,4′-DDOH

PESTANAL®, analytical standard

Synonym(s):

2,2-Bis(4-chlorophenyl)-1-ethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H12Cl2O
CAS Number:
Molecular Weight:
267.15
Beilstein:
2055080
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

SMILES string

OCC(c1ccc(Cl)cc1)c2ccc(Cl)cc2

InChI

1S/C14H12Cl2O/c15-12-5-1-10(2-6-12)14(9-17)11-3-7-13(16)8-4-11/h1-8,14,17H,9H2

InChI key

ZVIDYKRNLNAXFT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Eriksson et al.
Brain research, 514(1), 141-142 (1990-04-23)
DDT, 1,1,1-trichloro-2,2-bis(p-chlorophenyl)-ethane is one of the best-known insecticides which produces neurotoxic syndrome in mammals. DDT is also a world-wide environmental contaminant which is still used in several countries. We have previously reported on pronounced retention of DDT in the neonatal
Edward J Scollon et al.
Environmental toxicology and chemistry, 31(2), 336-346 (2011-11-03)
This study evaluated the interactions of flight, fasting, and 1,1,1-trichloro-bis(4-chlorophenyl)ethane (p,p'-DDT) loading on residue metabolism and distribution in recently exposed white-crowned sparrows (Zonotrichia leucophrys). Female sparrows were dosed with 5 mg p,p'-DDT per kg body weight over 3 d. Following

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service