Skip to Content
Merck
All Photos(1)

Key Documents

09989

Sigma-Aldrich

Zonyl® FSE fluorosurfactant

~14% solids in water: ethylene glycol (2.5:1)

Synonym(s):

anionic phosphate fluorosurfactant

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(RfCH2CH2O)xPO(ONH4)y(OCH2CH2OH)3-x-y
CAS Number:
MDL number:
UNSPSC Code:
12352300

Pricing and availability is not currently available.

concentration

~14% solids in water: ethylene glycol (2.5:1)

density

1.12 g/mL±0.05 g/mL at 20 °C

Legal Information

Zonyl is a registered trademark of E. I. du Pont de Nemours and Company

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service