Skip to Content
Merck
All Photos(1)

Key Documents

09730

Sigma-Aldrich

Ammonium iron(III) sulfate dodecahydrate

puriss. p.a., ACS reagent, ≥99.0% (RT)

Synonym(s):

Ammonium ferric sulfate dodecahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4Fe(SO4)2 · 12H2O
CAS Number:
Molecular Weight:
482.19
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

ACS reagent
puriss. p.a.

Assay

≥99.0% (RT)

mp

39-41 °C (lit.)

solubility

H2O: soluble 482.2 g/L at 20 °C

anion traces

chloride (Cl-): ≤5 mg/kg
nitrate (NO3-): ≤100 mg/kg
phosphate (PO43-): ≤30 mg/kg

cation traces

Ca: ≤20 mg/kg
Cd: ≤5 mg/kg
Co: ≤10 mg/kg
Cr: ≤50 mg/kg
Cu: ≤5 mg/kg
Fe2+: ≤10 mg/kg
K: ≤50 mg/kg
Mg: ≤10 mg/kg
Mn: ≤50 mg/kg
Na: ≤50 mg/kg
Ni: ≤10 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

N.O.O.O.O.O.O.O.O.O.O.O.O.[Fe+3].OS([O-])(=O)=O.[O-]S([O-])(=O)=O

InChI

1S/Fe.H3N.2H2O4S.12H2O/c;;2*1-5(2,3)4;;;;;;;;;;;;/h;1H3;2*(H2,1,2,3,4);12*1H2/q+3;;;;;;;;;;;;;;;/p-3

InChI key

LCPUDZUWZDSKMX-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Săndulescu et al.
Journal of pharmaceutical and biomedical analysis, 14(8-10), 951-958 (1996-06-01)
The method is based upon the reaction between fluoride ions and the coloured complex of Fe(III) with methyl salicylate to form the stable, colourless hexaflouride complex of iron. The conditions of the method (pH, time and combination ratio) were studied
S Puntarulo et al.
Archives of biochemistry and biophysics, 264(2), 482-491 (1988-08-01)
The interaction of microsomes with iron and NADPH to generate active oxygen radicals was determined by assaying for low level chemiluminescence. The ability of several ferric complexes to catalyze light emission was compared to their effect on microsomal lipid peroxidation
Luigi Brunetti et al.
Neurobiology of aging, 25(1), 125-129 (2003-12-17)
In order to investigate whether amyloid beta-peptide-induced oxidative damage in the brain could be related to aging, we studied the release of 8-iso-prostaglandin (PG)F2alpha, a stable marker of cellular oxidative stress, in brain synaptosomes from Wistar rats of different ages
J G Reinhold et al.
The Journal of nutrition, 116(6), 1007-1017 (1986-06-01)
The effects of pH, ascorbate, citrate and dietary fiber on retention of ferrous and ferric iron by jejuno-ileal segments of rat intestine were examined in vivo. Iron was introduced in an isosmotic solution of sodium chloride and dextrose buffered by
I Wittmann et al.
Chemico-biological interactions, 138(2), 171-187 (2001-10-24)
Diabetes mellitus is characterized by increased methylglyoxal (MG) production. The aim of the present study was to investigate the role of iron in the cellular and molecular effects of MG. A red blood cell (RBC) model and L-arginine were used

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service