Skip to Content
Merck
All Photos(1)

Key Documents

08941

Supelco

Citropten

analytical standard

Synonym(s):

5,7-Dimethoxycoumarin, Citropten, Limettin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H10O4
CAS Number:
Molecular Weight:
206.19
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

impurities

≤1.0% water

mp

146-149 °C (lit.)

application(s)

food and beverages

format

neat

SMILES string

COc1cc(OC)c2C=CC(=O)Oc2c1

InChI

1S/C11H10O4/c1-13-7-5-9(14-2)8-3-4-11(12)15-10(8)6-7/h3-6H,1-2H3

InChI key

NXJCRELRQHZBQA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Citropten is found in the essential oil of bergamot. It is a photomutagenic and phototoxic compound which is used to stimulate human skin pigmentation.[1]

Application

Citropten may be used as a reference standard in the determination of citropten in perfumes and sun-tan cosmetics using high performance liquid chromatography coupled with ultraviolet detector (HPLC-UV).[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High-performance liquid chromatographic determination of citropten and bergapten in suction blister fluid after solar product application in humans.
Makki S, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 563(2), 407-413 (1991)
Determination of bergapten and citropten in perfumes and suntan cosmetics by high-performance liquid chromatography and fluorescence.
Bettero A and Benassi CA
Journal of Chromatography A, 280(2), 167-171 (1983)
Adriana Arigò et al.
Journal of chromatography. A, 1649, 462183-462183 (2021-05-27)
In the European Union, cosmetic products are regulated by the Cosmetics Regulation (EC) No. 1223/2009. The Analytical Working Group of the International Fragrance Association (IFRA) suggested that a new sensitive analytical method is needed to determine psoralens in finished cosmetic
Yu Zhang et al.
Journal of pharmaceutical and biomedical analysis, 102, 110-118 (2014-09-30)
Spatholobi Caulis, the vine stem of Spatholobus suberectus Dunn, has been widely used in traditional Chinese and folk medicines for treatment of irregular menstruation, blood deficiency and rheumatalgia in clinic. In this study, an accurate and reliable high performance liquid
Raimo A Ketola et al.
Forensic science international, 249, e7-11 (2015-02-17)
Kavalactones are a group of compounds found in kava, a beverage or extract prepared from the rhizome of the kava plant (Piper methysticum). Traditionally kava extracts have been used for their anxiolytic and sedative properties. Sales of kava extracts were

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service