Skip to Content
Merck
All Photos(1)

Key Documents

07767

Sigma-Aldrich

N,N-Dimethyl-p-phenylenediamine dihydrochloride

≥99.5% (HPLC)

Synonym(s):

4-Amino-N,N-dimethylaniline dihydrochloride, DMPD · 2HCl, DMPPDA · 2HCl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NC6H4NH2 · 2HCl
CAS Number:
Molecular Weight:
209.12
Beilstein:
3913463
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

Assay

≥99.5% (HPLC)

mp

222 °C (dec.) (lit.)

anion traces

sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

Cl[H].Cl[H].CN(C)c1ccc(N)cc1

InChI

1S/C8H12N2.2ClH/c1-10(2)8-5-3-7(9)4-6-8;;/h3-6H,9H2,1-2H3;2*1H

InChI key

IAEDWDXMFDKWFU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Quantitation of labile sulfide in proteins by the "methylene blue" method; Detection of sterol hydroperoxides on irrigated thin-layer chromato-plates using N,N-dimethyl-p-phenylenediamine dihydrochloride; Preparation of the "nadi reagent" and demonstration of cytochrome oxidase activity in chick embryos

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of acid-labile sulfide and sulfhydryl groups.
J C Rabinowitz
Methods in enzymology, 53, 275-277 (1978-01-01)
F. Moog
Journal of Cellular and Comparative Physiology, 22, 223-223 (1943)
Detection of sterol hydroperoxides on thin-layer chromatoplates by means of the Wurster dyes.
L L Smith et al.
Journal of chromatography, 66(1), 101-109 (1972-03-22)
Veronica Verde et al.
Free radical research, 36(8), 869-873 (2002-11-08)
Oxidative stress has been clearly implicated in human disease by a growing body of scientific evidences. There is no ideal method for the measurement of this parameter. A possible strategy would be to measure simultaneously several biomarkers representing damage to
S Marroquí et al.
Journal of bacteriology, 183(3), 854-864 (2001-02-24)
We isolated a Tn5-induced Rhizobium tropici mutant that has enhanced capacity to oxidize N,N-dimethyl-p-phenylendiamine (DMPD) and therefore has enhanced respiration via cytochrome oxidase. The mutant had increased levels of the cytochromes c(1) and CycM and a small increase in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service