Skip to Content
Merck
All Photos(1)

Key Documents

05880595

Benzyl acetate

primary reference standard

Synonym(s):

Acetic acid benzyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOCH2C6H5
CAS Number:
Molecular Weight:
150.17
Beilstein:
1908121
EC Number:
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.24

grade

primary reference standard

vapor pressure

23 mmHg ( 110 °C)

autoignition temp.

862 °F

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

HWI

refractive index

n20/D 1.502 (lit.)

bp

206 °C (lit.)

mp

−51 °C (lit.)

density

1.054 g/mL at 25 °C (lit.)

application(s)

food and beverages

storage temp.

−20°C

SMILES string

CC(=O)OCc1ccccc1

InChI

1S/C9H10O2/c1-8(10)11-7-9-5-3-2-4-6-9/h2-6H,7H2,1H3

InChI key

QUKGYYKBILRGFE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.

Hazard Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

203.0 °F - closed cup

Flash Point(C)

95 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service