Skip to Content
Merck
All Photos(1)

Key Documents

11-0220

Sigma-Aldrich

Field′s reagent (B)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352106
form:
liquid
Pricing and availability is not currently available.

form

liquid

availability

available only in Japan

storage temp.

15-25°C

Suitability

blood stain

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service