Skip to Content
Merck
All Photos(1)

Key Documents

05-0720

Sigma-Aldrich

Calcium glycerophosphate hydrate

SAJ first grade, ≥98.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH(OH)CH2OPO3Ca
Molecular Weight:
210.14 (anhydrous basis)
UNSPSC Code:
12000000

grade

SAJ first grade

Assay

≥98.0%

form

solid

availability

available only in Japan

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service