Skip to Content
Merck
All Photos(1)

Documents

SX0255

Supelco

Sodium acetate trihydrate

Meets ACS Specifications GR ACS

Synonym(s):

Acetic Acid Sodium Salt, Acetic acid sodium salt trihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COONa · 3H2O
CAS Number:
Molecular Weight:
136.08
Beilstein:
3732037
MDL number:
UNSPSC Code:
12352107
EC Index Number:
204-823-8

Quality Level

Assay

99.0-101.0% (perchloric acid titration)

form

solid

autoignition temp.

1112 °F

impurities

≤0.005% Insoluble matter

pH

7.5-9.2 (20 °C, 50 g/L in H2O)

mp

58 °C

transition temp

flash point >250 °C (anhydrous substance)

solubility

soluble 613 g/L

anion traces

chloride (Cl-): ≤0.0005%
phosphate (PO43-): ≤0.0002%
sulfate (SO42-): ≤0.002%

cation traces

Ca: ≤0.001%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.0005%
heavy metals (as Pb): ≤0.0005%

shipped in

ambient

storage temp.

room temp

SMILES string

O.O.O.[Na+].CC([O-])=O

InChI

1S/C2H4O2.Na.3H2O/c1-2(3)4;;;;/h1H3,(H,3,4);;3*1H2/q;+1;;;/p-1

InChI key

AYRVGWHSXIMRAB-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Meets ACS Specifications GR ACS

Other Notes

Enzyme substrate for acetate kinase (EC 2.7.2.1)

Analysis Note

Assay (perchloric acid titration)99.0-101.0%Calcium (Ca)0.001% maxChloride (Cl)0.0005% maxHeavy metals (as Pb)0.0005% maxInsoluble matter0.005% maxIron (Fe)0.0005% maxMagnesium (Mg)0.0005% maxPhosphate (PO4)0.0002% maxPotassium (K)0.005% maxSubstances reducing permanganatepasses testSulfate (SO4)0.002% maxpH (5%, water)7.5-9.0%

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service