Skip to Content
Merck
All Photos(2)

Key Documents

NS830-M

Sigma-Aldrich

GFAP ELISA Kit

Synonym(s):

GFAP, GFAP, Glial Fibrillary Acidic Protein ELISA, Glial fibrillary acidic protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
Pricing and availability is not currently available.

species reactivity

rat, mouse, human

manufacturer/tradename

Chemicon®

assay range

sensitivity: 1.5 ng/mL
standard curve range: 1.5-100 ng/mL

technique(s)

ELISA: suitable

NCBI accession no.

UniProt accession no.

detection method

colorimetric

Gene Information

human ... GFAP(2670)

General description

Background
Glial Fibrillary Acidic Protein (GFAP) is a class-III intermediate filament uniquely and ubiquitously expressed as part of the structural network in astrocytic glial cells. This specific expression makes GFAP ideal as a broad astrocytic marker in vertebrates. Astrocytes play a variety of key roles in supporting, guiding, nurturing, and signaling neuronal architecture and activity in a highly interdependent manner. Current understanding of CNS structure suggests that there are at least as many glial cells as the estimated 100 billion neurons, but perhaps upwards of 100 times more. Thus,
the health of both neurons and astrocytes is often affected in disease and trauma. GFAP is proving to be a useful biomarker in the detection of neural degeneration. Although defects in GFAP have been reported to cause certain rare conditions, such as Alexander’s disease (Brenner et al., 2001), the observed interconnected physiology of neuronal and glial cells has enabled the use of GFAP as a neurodegeneration detection gauge. This relationship is being exploited by researchers and clinicians measuring a variety of neurological issues from cancer to trauma. GFAP has been used as a biomarker in the early detection of chemically induced cancer (Capo et al., 1997) and classical glioblastomas (Jung et al., 2007). Elevated levels of GFAP have been reported in traumatic brain injury (TBI) and appear to directly correlate with the severity of injury and brain tissue oxygenation in combat casualty care research (CCR, 2008) as well as civilian accidents (Mondello et al., 2010). GFAP has also been identified as a good biomarker in studies of toxin induced acute brain injury (Liao et al., 2008), in differentiation of hemorrhagic vs. ischemic strokes (Foerch et al., 2006), as well as in cardiac arrest (Kaneko et al., 2009). Millipore now offers an ELISA assay for sensitive measurement of GFAP protein in tissue homogenates and biological fluids (serum, plasma, CSF, and serum free media) from human, mouse, and rat.

Principals of Procedure
This assay is a Sandwich ELISA based sequentially on: 1) capture of soluble GFAP protein to the wells of a microtiter plate coated by a pre-titered amount of anti-GFAP monoclonal antibody; 2) removal of unbound material by washing; 3) binding of biotinylated anti-GFAP monoclonal antibody to
the previously captured GFAP protein; 4) removal of unbound material by washing; 5) binding of streptavidin conjugated horseradish peroxidase to the biotinylated antibodies; 6) removal of excess or unbound enzyme conjugates by washing; and 7) quantification of immobilized antibody-enzyme
complexes by monitoring horseradish peroxidase activity in the presence of the substrate 3,3’,5,5’-tetramethylbenzidine (TMB). Enzyme activity is measured spectrophotometrically by recording absorbance readings at 450 nm. Increases in absorbency are directly proportional to the amount of
GFAP protein captured from the sample. Therefore the protein concentration of unknowns can be derived by interpolation using a standard curve generated in the same assay with reference standards of known concentrations of GFAP.

Application

Research Category
Neuroscience
Used to detect/quantify: GFAP

Packaging

96 wells

Components

96-Well Anti-GFAP Coated Plate (CS206352): 1 Plate

1X Assay Buffer (CS206357): 40 mL

10X Wash Buffer (CS206360): 50 mL

GFAP Protein Standard (CS206361): 1 Vial

GFAP Quality Control #1 (CS206351): 1 Vial

GFAP Quality Control #2 (CS206475): 1 Vial

Biotinylated Anti-GFAP Detection Antibody (CS206356): 12 mL

Enzyme Solution (CS206358): 12 mL

TMB Solution (CS202682): 10 mL

Stop Solution (60193): 12 mL

Plate Sealers (CS200443): 10 Sheets

Storage and Stability

Storage: The GFAP ELISA kit is shipped on ice and should be stored at 2-8°C upon arrival.
Stability: Kit components are stable for a minimum of 6 months from the date of receipt if stored and
handled as described above.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

For research use only. Not for use in diagnostic procedures.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Met. Corr. 1 - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service