MCLHEAD01
Microfil® Head
Milliflex® Plus Pump, suitable for bioburden testing, water monitoring
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
material
stainless steel head
packaging
pkg of 1 unit
manufacturer/tradename
Milliflex®
input
sample type beverage(s)
sample type food(s)
application(s)
bioburden testing
water monitoring
clinical
environmental
food and beverages
industrial qc
compatibility
Milliflex® Plus Pump
shipped in
dry ice
General description
Microfil® filtration head is designed to improve ease of use, reduce maintenance and limit the risk of contamination. A continuous rim allows easy and safe membrane transfer with forceps, reducing the risk of touching the membrane filtration area. The porous filtration support can be removed simply by pressing down anywhere around its edge. These filtration heads offer full compatibility with the Milliflex® Plus Pump.
Application
Microfil® filtration head is an accessory of the Milliflex® Plus Pump used during filtration of the test samples for general microbiological analysis, bioburden testing, and water monitoring.
Legal Information
MICROFIL is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service