Skip to Content
Merck
All Photos(1)

Key Documents

MABN2529

Sigma-Aldrich

Anti-Huntingtin Disease (HD/HTT) Antibody

mouse monoclonal, MW8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

Product Name

Anti-HTT Antibody, clone MW8,

antibody form

purified antibody

clone

MW8
monoclonal

purified by

using protein G

species reactivity

mouse, mouse, human

concentration

(Please refer to lot specific datasheet.)

technique(s)

ELISA: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2aκ

UniProt accession no.

General description

HD exon-1 67Q aggregate.

Specificity

Clone MW8 is a mouse monoclonal antibody that detects Huntingtin protein. It targets an epitope within nine amino acids in the exon-1 region.

Application

Immunocytochemistry Analysis: A representative lot detected HTT in Immunocytochemistry applications (Legleiter, J., et. al. (2009). J Biol Chem. 284(32):21647-58).

Immunohistochemistry Analysis: A representative lot detected HTT in Immunohistochemistry applications (Ko, J., et. al. (2001). Brain Res Bull. 56(3-4):319-29).

Western Blotting Analysis: A representative lot detected HTT in Western Blotting applications (Legleiter, J., et. al. (2009). J Biol Chem. 284(32):21647-58).

Enzyme Immunoassay (ELISA) Analysis: A representative lot detected HTT in ELISA applications (Ko, J., et. al. (2001). Brain Res Bull. 56(3-4):319-29).

Immunoprecipitation Analysis: A representative lot immunoprecipitated HTT in Immunoprecipitation applications (Baldo, B., et. al. (2012). Chem Biol. 19(2):264-75).

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected HTT in Tissue lysate of 9 month-old WT or Q140 KI HD mice.

Quality

Isotype testing: Identity Confirmation by Isotyping Test.

Isotyping Analysis: The identity of this monoclonal antibody is confirmed by isotyping test to be mouse IgG2a.

Physical properties

347.6 kDa calclulated

Physical form

Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Human, Mouse.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service