Skip to Content
Merck
All Photos(1)

Documents

MAB5598

Sigma-Aldrich

Anti-HCN3 Antibody, clone TLL 6C5

culture supernatant, clone TLL 6C5, Chemicon®

Synonym(s):

Hyperpolarization-activated and cyclic nucleotide-gated channel 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

TLL 6C5, monoclonal

species reactivity

mouse, rat

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

mouse ... Hcn3(15168)

Specificity

HCN3 (Hyperpolarization-activated and cyclic nucleotide-gated channel 3). The antibody recognizes both the glycosyated and deglycosylated form of HCN3. The antibody recognizes a band at ~90 kDa in glycosylated protein from a retinal membrane preparation.

Immunogen

Synthetic peptide from rat HCN3.

Application

Anti-HCN3 Antibody, clone TLL 6C5 detects level of HCN3 & has been published & validated for use in IH & WB.
Western blot on rat and mouse retina. The antibody recognizes a band at ~90 kDa in glycosylated protein from a retinal membrane preparation.

Immunohistochemistry on rat and mouse retina.

Optimal working dilutions must be determined by end user.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nathan Grainger et al.
The Journal of physiology, 598(15), 3283-3307 (2020-05-18)
Platelet-derived growth factor receptor-α (PDGFRα) is a novel biomarker along with smooth myosin heavy chain for the pacemaker cells (previously termed 'atypical' smooth muscle cells) in the murine and cynomolgus monkey pelvis-kidney junction. PDGFRα+ cells present in adventitial and urothelial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service