Skip to Content
Merck
All Photos(1)

Documents

HTS156RTA

Sigma-Aldrich

Ready-to-Assay MC1 Melanocortin Receptor Frozen Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32011203
NACRES:
NA.76

description

Cell line type: GPCR Cell Lines
GPCR Class A
protein target MC1

Quality Level

technique(s)

cell based assay: suitable

detection method

fluorometric

General description

EMD Millipore′s Ready-to-Assay GPCR frozen cells are designed for simple, rapid calcium assays with no requirement for intensive cell culturing. EMD Millipore has optimized the freezing conditions to provide cells with high viability and functionality post-thaw. The user simply thaws the cells and resuspends them in media, dispenses cell suspension into assay plates and, following over night recovery, assays for calcium response.

The melanocortin-1 receptor (MC1) is a member of the G-protein-coupled receptor family of proteins that is responsible for regulating skin and hair color. The MC1 receptor is regulated by the physiological agonists alpha-melanocyte-stimulating hormone (α-MSH) and adrenocorticotropic hormone (ACTH), and antagonist agouti signaling protein (ASP) (Rouzud et al., 2005). In 1995 a landmark study demonstrated that over 80% of humans with red hair or fair skin have a dysfunctional variant of the MC1 gene (Valverde et al., 1995). MC1 is, therefore, a major determinant of sun sensitivity and genetic risk factor for melanoma and non-melanoma skin cancer. EMD Millipore′s cloned human MC1 receptor-expressing ChemiScreen cells were constructed by stable transfection of Chem-1 cells with the MC-1 receptor. These stability-tested cells are ready for cAMP accumulation assays for agonists, antagonists and modulators at the MC1 receptor.

Components

Pack contains 2 vials of mycoplasma-free cells, 1 ml per vial. Fifty (50) mL of Media Component.

Disclaimer

This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges {HCompany} to request certain information about you and the establishment where the GMOs are being handled. Click here for Enduser Declaration (EUD) Form.Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service