Skip to Content
Merck
All Photos(2)

Documents

E521-10014

Millipore

NovaSeptum® GO Transfer Unit

Tube Application; Luer; 500 mm; Si, sterile; β-irradiated

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104105

material

316 stainless steel cannula
C-Flex tubing
TPE tubing
polyester body
silicone septum (platinum-cured)
silicone tubing

Quality Level

Agency

according to ISO 11137 (sterilization)
according to ISO 146441
meets USP 88 Biological Reactivity (all component materials)

sterility

sterile; β-irradiated

product line

NovaSeptum® GO

feature

autoclavable: no

parameter

-50-95 °C temp. range (-58-203 °F)
0.50 bar max. pressure (7.25 psi)

impurities

<2.15 EU/device bacterial endotoxins (LAL test)

fitting

female Luer-Lok® outlet (3-piece)
male Luer-Lok® outlet (3-piece)
outlet (injection site)

application(s)

cell therapy
gene therapy
life science and biopharma
mAb
ophthalmics
parenterals
pharma/biopharma processes
sterile sampling
viral therapy

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

General description

Device configuration: single unit, Transfer tube set
The NovaSeptum® GO Transfer Unit is intended for transfer applications. It is available with a 2 mm needle and must be used with a NovaSeptum® GO holder. Transfer units are available with a range of different tubing materials (Thermoplastic, Silicone, and C-Flex®).

Application

Non-critical
Sampling

Packaging

500mm Silicone tube, 50 units per box

Components

  • Septum: Platinum-cured silicone
  • Body: Polyester
  • Cannula: ASTM® 316 L Stainless steel
  • Tubing: Thermoplastic elastomer (TPE), silicone or C-Flex® tube
  • Outlet Tubing: 3-piece Luer-Lok®, containing a male, female and an injection site

Preparation Note

Units are integrity tested at regular intervals during manufacturing.

Aqueous extraction contains < 2.15 EU per device as determined using the Limulus Amebocyte Lysate (LAL) test.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials
C-Flex is a registered trademark of Saint-Gobain Performance Plastics Corp.
Luer-Lok is a registered trademark of Becton-Dickinson & Co.
NOVASEPTUM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service