Skip to Content
Merck
All Photos(2)

Documents

ABS2053

Sigma-Aldrich

Anti-Glutaredoxin-2 (Grx2)

from rabbit, purified by affinity chromatography

Synonym(s):

Glutaredoxin-2, mitochondrial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

baboon (based on 100% sequence homology), monkey (based on 100% sequence homology), chimpanzee (based on 100% sequence homology)

packaging

antibody small pack of 25 μg

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... GLRX2(51022)

General description

Glutaredoxin-2, mitochondrial (UniProt: Q9NS18) is encoded by the GLRX2 (also known as GRX2, CGI-133) gene (Gene ID: 51022) in human. Glutaredoxin-2 is a glutathione-dependent oxidoreductase that is widely expressed and facilitates the maintenance of mitochondrial redox homeostasis upon induction of apoptosis by oxidative stress. Its active form is a monomer and it can dimerize to an inactive form where homodimers are linked by one 2Fe-2S cluster. The 2Fe-2S may serve as a redox sensor and the presence of one-electron oxidants or reductants can lead to the loss of the 2Fe-2S cluster and subsequent monomerization and activation of the enzyme. Glutaredoxin-2 acts as a very efficient catalyst of monothiol reactions because of its high affinity for protein glutathione-mixed disulfides. It can receive electrons not only from glutathione (GSH), but also from thioredoxin reductase supporting both monothiol and dithiol reactions. Glutaredoxin-2 is known to efficiently catalyze both glutathionylation and deglutathionylation of mitochondrial complex I, which in turn regulates the superoxide production by the complex. Overexpression of glutaredoxin-2 decreases the susceptibility to apoptosis and prevents loss of cardiolipin and cytochrome c release.

Specificity

This rabbit polyclonal antibody detects mitochondrial isoform of glutatedoxin-2. It targets an epitope with in 16 amino acids from the C-terminal half.

Immunogen

Epitope: unknown
KLH-conjugated linear peptide corresponding to 16 amino acids from the C-terminal half of human Glutaredoxin-2 .

Application

Anti-Glutaredoxin-2 (Grx2), Cat. No. ABS2053, is a rabbit polyclonal antibody that detects Glutaredoxin-2, mitochondrial, and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected Glutaredoxin-2 (Grx2) in human cerebellum and human tonsil tissue.
Research Category
Signaling

Quality

Evaluated by Western Blotting in A549 cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected Glutaredoxin-2 (Grx2) in 10 µg of A549 cell lysate.

Target description

~17 kDa observed; 18.05 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service