Skip to Content
Merck
All Photos(2)

Key Documents

AB2999

Sigma-Aldrich

Anti-BNIP3 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

BCL2/adenovirus E1B 19 kDa protein-interacting protein 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
polyclonal
application:
ICC
WB
species reactivity:
human
technique(s):
immunocytochemistry: suitable
western blot: suitable
citations:
2

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

mouse (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... BNIP3(664)

General description

BCL2/adenovirus E1B 19 kDa protein-interacting protein 3 (BNIP3) is a member of the NIP3 family, a singular group of death-inducing, mitochondrial proteins. BNIP3 is comprised of several domains of interest; a highly conserved domain (CD), a C-terminal transmembrane (TM) domain, and a BH3 domain. Strongly upregulated by hypoxia, BNIP3 induces a caspase-independent cell death characterized by mitochondrial damage. Overexpression has been observed in the brain after stroke and in the adult heart following myocardial infarction. Recent studies have shown BNIP3 to be involved in cell growth inhibition through suppression of the mTOR pathway and that, when localized to the nucleus, it has the paradoxical capability of repressing apoptosis in tumors by binding the AIF expression gene.

Specificity

Other homologies: Rat, equine, and ox (90% sequence homology).

Immunogen

Epitope: Internal (C-terminal half).
KLH-conjugated linear peptide corresponding to an internal sequence within the C-terminal half of human BNIP3.

Application

Anti-BNIP3 Antibody detects level of BNIP3 & has been validated for use in Immunocytochemistry and Western Blotting.
Immunocytochemistry Analysis: A representative lot detected hypoxia-induced BNIP3 upregulation in human glioblastoma-astrocytoma, epithelial-like U87 cells (Courtesy of Spencer Gibson, Manitoba Institute of Cell Biology).
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Quality

Evaluated by Western Blotting in PC3 cell lysates.

Western Blot Analysis: 0.5 µg/mL of this antibody detected BNIP3 in 10 µg of PC3 cell lysate.

Target description

~60 kDa (dimer) observed. BNIP3 may be observed as a 30 kDa monomer and/or a 60 kDa dimer.

Physical form

Affinity purified.
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
PC3 cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Yuan et al.
Cell death & disease, 4, e690-e690 (2013-06-29)
The histone methyltransferase G9a is overexpressed in a variety of cancer types, including pancreatic adenocarcinoma, and promotes tumor invasiveness and metastasis. We recently reported the discovery of BRD4770, a small-molecule inhibitor of G9a that induces senescence in PANC-1 cells. We
Lisa A Brennan et al.
Experimental eye research, 174, 173-184 (2018-06-08)
The formation and life-long growth of the ocular lens depends on the continuous differentiation of lens epithelial cells into lens fiber cells. To achieve their mature structure and transparent function, newly formed lens fiber cells undergo a series of cellular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service