Skip to Content
Merck
All Photos(1)

Key Documents

6.09992

Supelco

Titriplex® III solution

0.1 mol/l (0.1M), concentrate solution for titration, Titrisol®®

Synonym(s):

Complexometric titrant III

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352301

Product Name

Titriplex® III solution, for 500 ml, c(C10H14N2Na2O8.2H2O) = 0.1 mol/l (0.1M) Titrisol

form

solution

Application


  • The use of Titriplex III for cross-matching for urgent blood transfusions and patients with blood coagulation disorders: Demonstrates the application of Titriplex III in critical healthcare settings, particularly in ensuring the compatibility of blood transfusions, crucial for emergency medical procedures (Fischer & Busch, 1960).

  • Clinical use of ethylenediaminetetracetic acid, sodium salt (Titriplex III) as an anticoagulant: Highlights the role of Titriplex III in enhancing the safety and effectiveness of clinical practices by preventing blood coagulation during medical examinations and treatments (Dettori, 1958).

  • Calcium determination in blood with Titriplex III and indicator buffering tablets: This study showcases the utility of Titriplex III in biochemical analysis, specifically for the precise measurement of calcium levels in blood, an essential process in various diagnostic activities (Steber, 1956).

Legal Information

Titriplex is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service