Skip to Content
Merck
All Photos(1)

Key Documents

5.04293

Sigma-Aldrich

SRPK Inhibitor, SRPIN340

Synonym(s):

SRPK Inhibitor, SRPIN340, N-(2-Piperidin-1-yl-5-(trifluoromethyl)phenyl)isonicotinamide, Serine-Arginine-Rich Protein Kinase Inhibitor, SR Protein Kinase Inhibitor, SR Protein Phosphorylation Inhibitor 340, (SRPIN)340

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H18F3N3O
CAS Number:
Molecular Weight:
349.35
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77
Pricing and availability is not currently available.

Assay

≥99% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light beige

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

SMILES string

FC(F)(F)c1cc(c(cc1)N3CCCCC3)NC(=O)c2ccncc2

InChI

1S/C18H18F3N3O/c19-18(20,21)14-4-5-16(24-10-2-1-3-11-24)15(12-14)23-17(25)13-6-8-22-9-7-13/h4-9,12H,1-3,10-11H2,(H,23,25)

InChI key

DWFGGOFPIISJIT-UHFFFAOYSA-N

General description

A cell-permeable isonicotinamide that acts as an ATP-competitive SRPK1-selective inhibitor (IC50 = 0.14 and 1.8 µM, respectively, against mSRPK1 and mSRPK2) with much reduced activity against 143 other kinases. Shown to effectively counteract IGF-1-induced anti-angiogenic to pro-angiogenic VEGF isoforms switch both in cultures in vitro (1 h 10 µM SRPIN340 prior to 12 h IGF-1 stimulation of PCIPs) and in a murine hypoxia-induced retinal neovascularization model in vivo (10 pmol/µL/eye intraocular SRPIN340 injection) by inhibiting PKC/SRPK signaling-dependent, alternate splicing factor ASF- (SF2; splicing factor 2) mediated VEGF pro-mRNA PSS (proximal splice site) selection. Suppresses RNA virus Sindbis propagation (IC50 = 60 µM in Vero cultures) and HCV-JFH1 replication (1 & 10 µM SRPIN340 in Huh7.5.1 cultures). Exhibits no toxicity toward rats (2 g/kg p.o. for 2 wks), CHO (5 mg/mL for 24 h), or Huh7 (30 µM for 48 h).
AA cell-permeable isonicotinamide compound that acts as an ATP-competitive (Ki = 0.89 µM using mSRPK1), SRPK1-selective inhibitor (IC50 = 0.14 and 1.8 µM, respectively, against mSRPK1- or mSRPK2-catalyzed SF2 RS domain peptide phosphorylation) with much reduced or no activity against 143 other kinases, including Clk1 and Clk4, even at concentrations as high as 10 µM. Shown to effectively counteract IGF-1-induced anti-angiogenic to pro-angiogenic VEGF isoforms switch both in cultures in vitro (pro/anti VEGF mRNA ratio = 1.26- and 4.48-times of control ratio, respectively, in 12 h IGF-1 stimulated PCIPs with or without 1 h 10 µM SRPIN340 pretreatment) and in a murine hypoxia-induced retinal neovascularization model in vivo (Relative retinal VEGF mRNA content = 0.3 vs. 1.1, respectively, with or without 10 pmol/µL/eye intraocular SRPIN340 injection upon 48 h room air exposure of 6-day 75% O2-adopted P12 neonatal mice) by inhibiting PKC/SRPK signaling-dependent, alternate splicing factor ASF- (SF2; splicing factor 2) mediated VEGF pro-mRNA PSS (proximal splice site) selection. Suppresses RNA virus Sindbis propagation (IC50 = 60 µM as determined by virus titre in 4 d-infected Vero cultures) and HCV-JFH1 replication (% HCV core protein-positive Huh7.5.1 48 h post infection = 18.2, 5.9, and 3.0, respectively, with 0. 1. 10 µM SRPIN340; MOI = 0.1). Exhibits no mutagenic effects by Salmonella typhimurium AMES test, nor toxicity toward rats (2 g/kg p.o. for 2 wks), CHO (5 mg/mL for 24 h), or Huh7 (30 µM for 48 h).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Karakama, Y., et al. 2010. Antimicrob. Agents Chemother.54, 3179.
Nowak, D.G., et al. 2010. J. Biol. Chem.285, 5532.
Fukuhara, T., et al. 2006. Proc. Natl. Acad. Sci. USA103, 11329.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service