Skip to Content
Merck
All Photos(1)

Documents

444153

Sigma-Aldrich

MDM2/MDMX Dual Inhibitor, RO-5963

Synonym(s):

MDM2/MDMX Dual Inhibitor, RO-5963, MDM2 Inhibitor VIII, MDMX Inhibitor III, ( Z)-2-(4-((6-Chloro-7-methyl-1H-indol-3-yl)methylene)-2,5-dioxoimidazolidin-1-yl)-2-(3,4-difluorophenyl)-N-(1,3-dihydroxypropan-2-yl)acetamide, MDM2 Antagonist V, MDM2 Inhibitor VIII, MDMX Inhibitor III, (Z)-2-(4-((6-Chloro-7-methyl-1H-indol-3-yl)methylene)-2,5-dioxoimidazolidin-1-yl)-2-(3,4-difluorophenyl)-N-(1,3-dihydroxypropan-2-yl)acetamide, MDM2 Antagonist V

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H21ClF2N4O5
Molecular Weight:
518.90
UNSPSC Code:
12352200

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

orange-brown

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A cell-permeable indolyl-hydantoin compound that simultaneously inhibits the interaction of both, MDM2 and MDMX with p53 (IC50 = 17.3 and 24.7 nM, respectively, in vitro). Releases p53 from MDMX inhibition, stabilizes it, and fully restores its function. Shown to be far more superior to Nutlin-3a (>Cat. No. 444152) in blocking p53 interaction with MDMX. Binds to Phe, Trp, and Leu sub-pockets on the surface of MDMX and MDM2, and promotes MDMX and MDM2 dimer formation. Shown to induce cell cycle arrest in G1 and G2 phases. Also reported to induce apoptosis in several tumor cell lines expressing wild-type p53/MDMX (~ 10 to 20 µM). Its actions are not believed to be due to any genotoxic stress.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
MDMX
Reversible: yes
Secondary Target
MDM2

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Graves, B., et al. 2012. Proc. Natl. Acad. Sci. USA109, 11788.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service