Skip to Content
Merck
All Photos(2)

Key Documents

420203

Sigma-Aldrich

JMJD Histone Demethylase Inhibitor IV, Methylstat

The JMJD Histone Demethylase Inhibitor IV, Methylstat controls the biological activity of JMJD Histone Demethylase. This small molecule/inhibitor is primarily used for Cell Structure applications.

Synonym(s):

JMJD Histone Demethylase Inhibitor IV, Methylstat, JHDM Inhibitor IV, ( E)-Methyl-4-(hydroxy(4-((4-(((naphthalen-1-ylcarbamoyl)oxy)methyl)benzyl)amino)butyl)amino)-4-oxobut-2-enoate, Histone Lysine Demethylase Inhibitor VII, JHDM Inhibitor IV, (E)-Methyl-4-(hydroxy(4-((4-(((naphthalen-1-ylcarbamoyl)oxy)methyl)benzyl)amino)butyl)amino)-4-oxobut-2-enoate, Histone Lysine Demethylase Inhibitor VII

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H31N3O6
CAS Number:
Molecular Weight:
505.56
MDL number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.

Quality Level

Assay

≥96% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A methyl ester prodrug of in vitro active N-hydroxybutenoic acid compound (Cat. No. 420202) that is shown to preferentially induce hypermethylation of histone proteins (EC50 for H3K4me3 and H3K9me3 = 10.3 and 8.6 µM in KYSE150 cells, and 6.7 and 6.3 µM in MCF7 cells, respectively), and arrests cell growth. Acts as a selective inhibitor of Jumonji domain-containing histone demethylases (JHDMs) over prolyl hydroxylases (PHDs), and prevents myogenesis through inhibition of H3K27 demethylase UTX in C2C12 cells at 2 µM.
A methyl ester prodrug of in vitro active N-hydroxybutenoic acid compound (Cat. No. 420202) that is shown to preferentially induce hypermethylation of histone proteins (EC50 for H3K4me3 and H3K9me3 = 10.3 and 8.6 µM in KYSE150 cells, and 6.7 and 6.3 µM in MCF7 cells, respectively), and arrests cell growth. Acts as a selective inhibitor of Jumonji domain-containing histone demethylases (JHDMs) over prolyl hydroxylases (PHDs), and prevents myogenesis through inhibition of H3K27 demethylase UTX in C2C12 cells at 2 µM.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Luo, X., et al. 2011. J. Am. Chem. Soc.133, 9451.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service