Skip to Content
Merck
All Photos(1)

Documents

1.50208

Supelco

LiChrospher® RP-4 ADS LiChroCART® 25-4

pkg of 3 cartridges

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115709

product line

LiChrospher®

Quality Level

packaging

pkg of 3 cartridges

technique(s)

solid phase extraction (SPE): suitable

storage temp.

no temp limit

General description

3 HPLC cartridges
LiChrospher® ADS allows the fully automated, LC integrated sample preparation of hydrophobic, low molecular weight analytes from untreated samples like serum, milk, fermentation broth of food homogenates. The enrichment is based on two chromatographic processes: reversed phase and size exclusion chromatography since LiChrospher ADS sorbents are spherical silica gel particles (25µm) with DIOL modification on the exterior surface and alkyl modifications on the surface of the interior pores (ID: 6nm). Depending on the hydrophobicity of the analyte three types of precolumns are available: C-4, C-8 and C-18.

Analysis Note

Pressure: ≤ 5 bar
Theoretical Plates (N/m) (Toluene): ≥ 3000
Symmetry (Toluene): 0.8 - 1.8
Capacity factor (Toluene): ≥ 0.3

Legal Information

LICHROCART is a registered trademark of Merck KGaA, Darmstadt, Germany
LiChrospher is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service