Skip to Content
Merck
All Photos(1)

Documents

1.50207

Supelco

LiChrospher® RP-8 ADS LiChroCART® 25-4

Sign Into View Organizational & Contract Pricing


About This Item

product line

LiChrospher®

Quality Level

technique(s)

solid phase extraction (SPE): suitable

General description

HPLC cartridge set, contains 1 LiChrospher® RP-8 ADS LiChroCART® 25-4 and 1 manu-CART®
LiChrospher® ADS allows the fully automated, LC integrated sample preparation of hydrophobic, low molecular weight analytes from untreated samples like serum, milk, fermentation broth of food homogenates. The enrichment is based on two chromatographic processes: reversed phase and size exclusion chromatography since LiChrospher® ADS sorbents are spherical silica gel particles (25μm) with  DIOL modification on the exterior surface and alkyl modifications on the surface of the interior pores (ID: 6nm). Depending on the hydrophobicity of the analyte three types of precolumns are available: C-4, C-8 and C-18.

Storage and Stability

Storage temperature: no restrictions.

Analysis Note

Extent of testing:passes test

Legal Information

LICHROCART is a registered trademark of Merck KGaA, Darmstadt, Germany
LiChrospher is a registered trademark of Merck KGaA, Darmstadt, Germany
MANU-CART is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service