Skip to Content
Merck
All Photos(1)

Key Documents

07-1520

Sigma-Aldrich

Anti-phospho-Grb10 (Ser501/Ser503) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

growth factor receptor-bound protein 10, OTTHUMP00000197988, OTTHUMP00000197989, OTTHUMP00000197990, OTTHUMP00000197991, OTTHUMP00000197992, OTTHUMP00000197993, OTTHUMP00000197994, GRB10 adapter protein, GRB10 adaptor protein, maternally expressed gene 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, horse, pig

species reactivity (predicted by homology)

rhesus macaque (based on 100% sequence homology), equine (based on 100% sequence homology), primate (based on 100% sequence homology), porcine (based on 100% sequence homology), human (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pSer501/pSer503)

Gene Information

human ... GRB10(2887)

General description

Growth factor receptor-bound protein 10 (Grb10) belongs to a small family of structurally related multi-domain adaptor proteins, which are known to be involved with a diverse array of functions, including cellular growth, metabolism, apoptosis, and cell migration. Grb10 is widely, but not uniformly, expressed in mammalian tissues, and there are various known isoforms of Grb10. It is shown to bind to and positively or negatively mediate signals from a number of activated receptor tyrosine kinases, including the insulin (INSR) and the insulin-like growth factor (IGF1R and IGF2R) receptors, in addition to a variety of growth factor receptors and intracellular molecules, in a context-dependent manner. In one study, mTORC1-mediated Grb10 phosphorylation is demonstrated to lead to feedback inhibition of the phosphatidylinositol-3-kinase (PI3K) and the extracellular signal-regulated, mitogen-activated protein kinase (ERK-MAPK) pathways in HEK 293T cells, in vivo. However, many aspects of the Grb10 adapter function remain unclear. Further research is still required to elucidate and validate the interactions and effects of GRB10 with various cellular targets.

Specificity

Other homologies: Rat (91% sequence homology).
This antibody recognizes Grb10 phosphorylated at Ser501/Ser503.

Immunogen

Epitope: Phosphorylated Ser501/Ser503.
KLH-conjugated linear peptide corresponding to human Grb10 phosphorylated at Ser501/Ser503.

Application

Anti-phospho-Grb10 (Ser501/Ser503) Antibody detects level of phospho-Grb10 (Ser501/Ser503) & has been published & validated for use in WB.
Research Category
Metabolism
Research Sub Category
Glucose/Glycogen Metabolism

Quality

Evaluated by Western Blot in insulin untreated and treated mouse embryonic fibroblast cell lysates.

Western Blot Analysis: 1 µg/mL of this antibody detected Grb10 in 10 µg of insulin untreated and treated mouse embryonic fibroblast cell lysates.

Target description

~66 kDa observed. Uncharacterized bands detected in some lysates. UniProt describes 4 isoforms produced by alternative splicing: Isoform 1 at 62.026 kDa, Isoform 2 at 60.848 kDa, Isoform 3 at 67.231 kDa, and Isoform 4 at 66.487 kDa

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Insulin untreated and treated mouse embryonic fibroblast cell lysates

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Agus Suryawan et al.
The Journal of nutrition, 148(6), 825-833 (2018-05-26)
Feeding stimulates protein synthesis in skeletal muscle of neonates and this response is regulated through activation of mechanistic target of rapamycin complex 1 (mTORC1). The identity of signaling components that regulate mTORC1 activation in neonatal muscle has not been fully
Phosphoproteomic analysis identifies Grb10 as an mTORC1 substrate that negatively regulates insulin signaling.
Yu, Y; Yoon, SO; Poulogiannis, G; Yang, Q; Ma, XM; Villen, J; Kubica, N; Hoffman et al.
Science (New York, N.Y.) null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service