Skip to Content
Merck
All Photos(1)

Documents

Z699187

MI-Broth Media

pack of 50 × 2 mL

Synonym(s):

e coli media, media broth

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NB.79

packaging

pack of 50 × 2 mL

manufacturer/tradename

Maine Mfg 10496192

application(s)

food and beverages

General description

MI Broth and MI Agar
MI Broth detects the presence of coliform bacteria by the production of b-galactosidase, which cleaves the substrate MUGal to produce 4-methylumbelliferone, which fluoresces on exposure to UV light. Non-coliforms do not produce this enzyme and therefore do not fluoresce on the medium. Escherichia coli is detected by the compound IBDG. The b-glucuronidase produced by Escherichia coli cleaves the substrate to produce a blue indigo color in the colonies. As Escherichia coli is also a total coliform, and also produces b-galactosidase, it will also fluoresce. The antibiotic cefsulodin is present to inhibit the growth of gram-positive bacteria and some non-coliform gram-negative bacteria that can cause false positive reactions.

Other Notes

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service