Recommended Products
AC/DC input
230 V
manufacturer/tradename
IKA 2621900
availability
not available in USA and Canada
parameter
0-1000 rpm speed
0.1 nm max. torque
H × W × D
110 mm × 360 mm × 430 mm
platform capacity
70000 g
stirrer capacity
50 L
weight
9 kg
Looking for similar products? Visit Product Comparison Guide
General description
These powerful magnetic stirrers feature digital LED speed/timer displays and 304 SS casings.
Legal Information
IKAMAG is a trademark of IKA-Werke GmbH & Co. KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service