Documents
Z189944
Sigma-Aldrich
ACE heavy-wall powder funnels
Straight, top diam. 75 mm
About This Item
UNSPSC Code:
41121809
NACRES:
NB.43
Joint
joint ST/NS24/40
manufacturer/tradename
Ace Glass 723505
top diam.
75 mm
Related Categories
General description
For pouring powders or liquids into ground-joint containers.