Documents
Z112151
Sigma-Aldrich
32mm Round cell window
NaCl, Drilled
About This Item
UNSPSC Code:
12000000
NACRES:
SB.32
material
NaCl
packaging
pkg of (contains 1 window)
technique(s)
IR spectroscopy: suitable
diam. × thickness
32 mm × 3 mm
General description
Cut and polished for demountable liquid and gas cell holders