Skip to Content
Merck
All Photos(1)

Key Documents

W318604

Sigma-Aldrich

4-Phenyltoluene

≥98%

Synonym(s):

4-Methylbiphenyl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H4CH3
CAS Number:
Molecular Weight:
168.23
FEMA Number:
3186
EC Number:
Council of Europe no.:
2292
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
1.011
Organoleptic:
spicy; floral; wintergreen
grade:
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

Kosher

Assay

≥98%

bp

267-268 °C (lit.)

mp

44-47 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

spicy; floral; wintergreen

SMILES string

Cc1ccc(cc1)-c2ccccc2

InChI

1S/C13H12/c1-11-7-9-13(10-8-11)12-5-3-2-4-6-12/h2-10H,1H3

InChI key

ZZLCFHIKESPLTH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Taste at 12 ppm

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mosciano, G.
Perfumer & Flavorist, 21, 53-53 (1996)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service