MT51343050
Mettler-Toledo InLab® Routine & Routine Pro pH electrodes
L × diam. 120 mm × 12 mm, S7 connector
Synonym(s):
combination pH electrode with S7 connector
About This Item
Recommended Products
material
glass shaft
glass, HA membrane
description
Reference electrolyte 3 mol/L KCl
Reference system ARGENTHAL™ with Ag+ -trap
Without cable
feature
S7 connector
manufacturer/tradename
Mettler-Toledo 51343050
temperature measuring range
0-100 °C
L × diam.
120 mm × 12 mm
pH range
0-14
Looking for similar products? Visit Product Comparison Guide
General description
Designed for accurate measurement in a wide range of sample types.
The electrolyte in conventional silver/silver chloride reference systems must be saturated with silver chloride. With critical media such as those containing sulphides or proteins, this will result in diaphragm contamination with silver sulphide. This cannot happen with InLab electrodes which have an ARGENTHAL™ lead off and silver ion trap that ensure the electrolyte remains completely free from silver ions.
The ARGENTHAL reference system also improves service life and measurement accuracy at differing sample temperatures. The glass shaft provides resistance against organic and corrosive samples.
Different cables can be connected to the S7 screw head allowing use with a wide range of meters.
- ARGENTHAL reference
- Silver ion trap
- Glass shaft
- Detachable cable
Legal Information
related product
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service