Skip to Content
Merck
All Photos(1)

Documents

L511811

Sigma-Aldrich

9-Mesityl-10-methylacridinium perchlorate

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H22ClNO4
CAS Number:
Molecular Weight:
411.88
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

SMILES string

CC1=CC(C)=CC(C)=C1C2=C3C(C=CC=C3)=[N+](C)C4=CC=CC=C42.O=Cl(=O)([O-])=O

InChI

1S/C23H22N.ClHO4/c1-15-13-16(2)22(17(3)14-15)23-18-9-5-7-11-20(18)24(4)21-12-8-6-10-19(21)23;2-1(3,4)5/h5-14H,1-4H3;(H,2,3,4,5)/q+1;/p-1

InChI key

LFMBERYWDLWXNO-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Metal-free, visible-light mediated catalyst used by Nicewicz (BF4 analog) for anti-Markovnikov hydroamination and hydroetherification reactions.

Anti-Markovnikov Hydroamination of Alkenes Catalyzed by an Organic Photoredox System

Used by Griesbeck as a visible-light mediated oxygenation catalyst in the presence of O2.

9-Mesityl-10-methylacridinium: An Efficient Type II and Electron-Transfer Photooxygenation Catalyst

For other Visible-Light Photocatalysts, refer to: www.aldrich.com/photocatalysis

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Pictograms

Flame over circleExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service