Skip to Content
Merck
All Photos(1)

Key Documents

GF63683804

Nickel silver alloy, Cu62Ni18Zn20

100mm tube, 1.28mm outside diameter, 0.120mm wall thickness, 1.04mm inside diameter, as drawn

Synonym(s):

CU107160

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11101711
NACRES:
NA.23

form

tubes

manufacturer/tradename

Goodfellow 636-838-04

L × wall thickness

100 mm × 0.120 mm

O.D. × I.D.

1.28 mm × 1.04 mm

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service