D155500
Dimethyl 2,4-dimethylglutarate
98%, Mixture of (±) and meso
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3O2CCH(CH3)CH2CH(CH3)CO2CH3
CAS Number:
Molecular Weight:
188.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Assay
98%
refractive index
n20/D 1.425 (lit.)
density
1.037 g/mL at 25 °C (lit.)
SMILES string
COC(=O)C(C)CC(C)C(=O)OC
InChI
1S/C9H16O4/c1-6(8(10)12-3)5-7(2)9(11)13-4/h6-7H,5H2,1-4H3
InChI key
LQLYZPTVIYLWPB-UHFFFAOYSA-N
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
201.2 °F - closed cup
Flash Point(C)
94 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service