Skip to Content
Merck
All Photos(1)

Key Documents

A91000

Sigma-Aldrich

Anthrarobin

technical grade, 85%

Synonym(s):

1,2,10-Anthracenetriol, Desoxyalizarin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H10O3
CAS Number:
Molecular Weight:
226.23
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

grade

technical grade

Assay

85%

mp

218 °C (dec.) (lit.)

SMILES string

Oc1ccc2c(O)c3ccccc3cc2c1O

InChI

1S/C14H10O3/c15-12-6-5-10-11(14(12)17)7-8-3-1-2-4-9(8)13(10)16/h1-7,15-17H

InChI key

TZIQWQARHPGHIG-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Allergic contact dermatitis from anthrarobin.
R Keller-Melchior et al.
Contact dermatitis, 33(5), 361-361 (1995-11-01)
A Krebs et al.
The British journal of dermatology, 105 Suppl 20, 6-11 (1981-08-01)
In the investigations of Krebs and Schaltegger between 1964 and 1972 on the structure specificity of the antipsoriatic anthrones it could be shown that at least a 1-hydroxy-9-anthrone is necessary for their efficacy. This structure was then called "minimum structure

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service