Skip to Content
Merck
All Photos(1)

Documents

902594

Sigma-Aldrich

Diisopropyl 2-oxomalonate

≥95%

Synonym(s):

Di-isopropyl ketomalonate, Kurti amination reagent, Kurti amine umpolung reagent

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H14O5
CAS Number:
Molecular Weight:
202.20
UNSPSC Code:
12352101

Assay

≥95%

form

liquid

refractive index

n/D 1.414

density

1.036

InChI

1S/C9H14O5/c1-5(2)13-8(11)7(10)9(12)14-6(3)4/h5-6H,1-4H3

InChI key

ISAKWFYKUTYAQE-UHFFFAOYSA-N

Application

As demonstrated by Laszlo Kürti′s lab, the sterically hindered di-isopropyl keto malonate and its hydrate (902764) are excellent N-umpolung reagents for both aliphatic and aromatic primary amines via a simple condensation reaction. The steric hindrance in ester moiety allows the preparation of the corresponding iminomalonates in high yield and lends these imines sufficient hydrolytic stability during the purification process. The N-alkyl as well as N-aryl iminomalonates act as singly N-electrophilic reagents towards strong C-nucleophiles such as alkyl- and arylmetals (e.g., Grignard reagents). Thus, unsymmetrical secondary amines may be readily prepared at low temperatures and in the absence of transition metal catalysts.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dialkyl mesoxalates by ozonolysis of dialkyl benzalmalonates: preparation of dimethyl mesoxalate.
Organic Syntheses, 71 (1993)
Facile synthesis of 1,2,3-tricarbonyls from 1,3-dicarbonyls mediated by cerium(IV) ammonium nitrate.
Sivan A, et al.
Tetrahedron Letters, 55, 1890-1893 (2014)
Intramolecular electrophilic cyclization of double activated imines induced by Lewis acids and trialkylsilyl triflates. An efficient route to substituted piperidines and annulated piperidine lactones.
Tietze L F, et al.
European Journal of Organic Chemistry, 122, 997-1002 (1989)
Ying Zhi et al.
Chemical communications (Cambridge, England), 52(97), 14011-14014 (2016-11-18)
The asymmetric synthesis of highly functionalized pyrrolidine derivatives with three contiguous stereogenic centers and bearing a trifluoromethyl group has been developed through an organocatalytic domino Michael/Mannich [3+2] cycloaddition sequence. Employing a commercially available secondary amine as the catalyst, the scalable
Electrophilic cyclization of doubly activated imines with Lewis acids and trialkylsilyl triflates. Synthesis of pyrrole and azepine derivatives.
Tietze L F, et al.
European Journal of Organic Chemistry, 6, 559-564 (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service