Skip to Content
Merck
All Photos(1)

Key Documents

900668

Sigma-Aldrich

(4-Hydroxyphenyl)methacrylamide

≥98%

Synonym(s):

N-(4-Hydroxyphenyl)-2-methyl-2-propenamide, N-(4-Hydroxyphenyl)methacrylamide, N-(p-Hydroxyphenyl)methacrylamide, p-Hydroxymethacrylanilide, Acrylanilide, 4′-hydroxy-2-methyl-(7CI,8CI)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11NO2
CAS Number:
Molecular Weight:
177.20
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.23

Assay

≥98%

form

powder

mp

142-147 °C

storage temp.

2-8°C

InChI

1S/C10H11NO2/c1-7(2)10(13)11-8-3-5-9(12)6-4-8/h3-6,12H,1H2,2H3,(H,11,13)

InChI key

XZSZONUJSGDIFI-UHFFFAOYSA-N

Application

Monomer for biomedical and lithographic applications. May be used to make polymer substrates that support cell expansion and differentiation.[1]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adam D Celiz et al.
Advanced materials (Deerfield Beach, Fla.), 27(27), 4006-4012 (2015-06-03)
A scalable and cost-effective synthetic polymer substrate that supports robust expansion and subsequent multilineage differentiation of human pluripotent stem cells (hPSCs) with defined commercial media is presented. This substrate can be applied to common cultureware and used off-the-shelf after long-term

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service