Skip to Content
Merck
All Photos(1)

Key Documents

804347

Sigma-Aldrich

3-nitro-N-[(3S)-tetrahydro-2-oxo-furanyl]-Benzeneacetamide

Synonym(s):

LasR inhibitor, LuxR activator

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12N2O5
CAS Number:
Molecular Weight:
264.23
UNSPSC Code:
12352200
NACRES:
NA.22

form

solid

storage temp.

−20°C

Application

PHL has varied activity depending on species, for example serving as either a strong LuxR-type receptor activator (e.g.: of LuxR in V. fischeri and of ExpR1 and ExpR2 in Pectobacterium carotovora[1]) or a strong inhibitor (e.g.: of LasR in P. aeruginosa[2])

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrew G Palmer et al.
Chembiochem : a European journal of chemical biology, 12(1), 138-147 (2010-12-15)
Many bacteria use quorum sensing (QS) to regulate cell-density dependent phenotypes that play critical roles in the maintenance of their associations with eukaryotic hosts. In Gram-negative bacteria, QS is primarily controlled by N-acylated L-homoserine lactone (AHL) signals and their cognate
Grant D Geske et al.
Journal of the American Chemical Society, 129(44), 13613-13625 (2007-10-12)
Bacteria use a language of low molecular weight ligands to assess their population densities in a process called quorum sensing. This chemical signaling process plays a pivotal role both in the pathogenesis of infectious disease and in beneficial symbioses. There

Related Content

We study quorum sensing, how bacteria sense their density. Synthesized compounds modulate pathways, aiding research on infections in various hosts.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service