Skip to Content
Merck
All Photos(1)

Key Documents

797413

Sigma-Aldrich

2-(tert-Butoxycarbonyl)-5-oxa-2-azaspiro[3.4]octane-7-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H19NO5
CAS Number:
Molecular Weight:
257.28
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

solid

functional group

carboxylic acid
ether

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(N1CC2(CC(CO2)C(O)=O)C1)=O

InChI

1S/C12H19NO5/c1-11(2,3)18-10(16)13-6-12(7-13)4-8(5-17-12)9(14)15/h8H,4-7H2,1-3H3,(H,14,15)

InChI key

YQIOAEQQZQISNG-UHFFFAOYSA-N

Application

Spirocyclic building block was first synthesized by Carreira and coworkers, which provides a new area of chemical space with straightforward functional handles for further diversification.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Spirocyclic modules containing four-membered rings are currently of growing interest to discovery chemists.

Related Content

Carreira Research & SpiroChem innovate spirocyclic building blocks, enhancing drug discovery with diverse functionalization sites and compact modules.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service