Skip to Content
Merck
All Photos(2)

Documents

752967

Sigma-Aldrich

Phenyltriethylammonium iodide

97%

Synonym(s):

Triethylphenylammonium iodide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H20IN
CAS Number:
Molecular Weight:
305.20
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

127-133 °C

SMILES string

[I-].CC[N+](CC)(CC)c1ccccc1

InChI

1S/C12H20N.HI/c1-4-13(5-2,6-3)12-10-8-7-9-11-12;/h7-11H,4-6H2,1-3H3;1H/q+1;/p-1

InChI key

WMSWXWGJYOIACA-UHFFFAOYSA-M

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G L Zhang et al.
Yao xue xue bao = Acta pharmaceutica Sinica, 25(8), 604-607 (1990-01-01)
Six ingredients were isolated from the whole plant of Dactylicapnos torulosa (Hook. f. et Thomas.) Hutchins. Four of them were identified by UV, IR, 1HNMR and 13CNMR. One of them is a new compound named zijinlongine(I). The three others are
Neal Whitaker et al.
Journal of pharmaceutical sciences, 106(11), 3230-3241 (2017-07-03)
High protein concentration formulations are required for low-volume administration of therapeutic antibodies targeted for subcutaneous, self-administration by patients. Ultra-high concentrations (≥150 mg/mL) can lead to dramatically increased solution viscosities, which in turn can lead to stability, manufacturing, and delivery challenges. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service