Skip to Content
Merck
All Photos(1)

Documents

749060

Sigma-Aldrich

Zinc

sputtering target, diam. × thickness 3.00 in. × 0.125 in., 99.995% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Zn
CAS Number:
Molecular Weight:
65.39
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

1 mmHg ( 487 °C)

Assay

99.995% trace metals basis

form

solid

resistivity

5.8 μΩ-cm, 20°C

diam. × thickness

3.00 in. × 0.125 in.

bp

907 °C (lit.)

mp

420 °C (lit.)

density

7.133 g/mL at 25 °C (lit.)

SMILES string

[Zn]

InChI

1S/Zn

InChI key

HCHKCACWOHOZIP-UHFFFAOYSA-N

Application

Sputtering is a process whereby atoms are ejected from a solid target material due to bombardment of the target by energetic particles. It is commonly used for thin-film deposition, etching and analytical techniques.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hahn, H.; Averback, R. S.
Journal of Applied Physics, 67, 1113-1113 (1990)
Dahn, J. R.; Trussler, S.; Hatchard, T. D.; Bonakdarpour, B. et al.
Chemistry of Materials, 14, 3519-3519 (2002)
Sigmund, P.
Physical Review, 184, 383-383 (1969)
Oechsner, H.
Applied Physics. A, Materials Science & Processing, 8, 185-185 (1975)
Sara Eyal et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 50(5), 798-806 (2009-05-01)
Studies in rodents indicate that the disruption of P-glycoprotein (P-gp) function increases drug distribution into the developing fetus and organs such as the brain. To simultaneously and serially evaluate the effect of P-gp activity and inhibition on the tissue distribution

Articles

Nanocomposite Coatings with Tunable Properties Prepared by Atomic Layer Deposition

Continuous efficiency improvements in photovoltaic devices result from material advancements and manufacturing innovation.

Spintronics offer breakthroughs over conventional memory/logic devices with lower power, leakage, saturation, and complexity.

Combinatorial Materials Science identifies breakthrough materials through systematic exploration, aiding material discovery.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service