Skip to Content
Merck
All Photos(1)

Key Documents

733415

Sigma-Aldrich

Poly(ethylene glycol) ditosylate

average Mn 20,000

Synonym(s):

PEG ditosylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4SO2(OCH2CH2)nOSO2C6H4CH3
UNSPSC Code:
12162002
NACRES:
NA.23

form

solid

mol wt

average Mn 20,000

transition temp

Tm 55-62 °C

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service