Skip to Content
Merck
All Photos(1)

Key Documents

679879

Sigma-Aldrich

S,S-Dimethyl N-(2,2,2-trichloroethoxysulfonyl)carbonimidodithionate

97%

Synonym(s):

[(2,2,2-Trichloroethoxy)sulfonyl)]-carbonimidodithionic acid dimethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H8Cl3NO3S3
CAS Number:
Molecular Weight:
332.68
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

69-73 °C

storage temp.

2-8°C

SMILES string

CS\C(SC)=N/S(=O)(=O)OCC(Cl)(Cl)Cl

InChI

1S/C5H8Cl3NO3S3/c1-13-4(14-2)9-15(10,11)12-3-5(6,7)8/h3H2,1-2H3

InChI key

YLVMQHRVOAWREM-UHFFFAOYSA-N

Application

  • Reactant for oxidative cyclization reactions
Reagent for Rh-catalyzed guanidine formation.
Starting material for preparation of guanidination reagents.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Editorial 2006: exciting and challenging times for organic letters.
Amos B Smith
Organic letters, 8(1), 1-2 (2005-12-31)
Mihyong Kim et al.
Organic letters, 8(6), 1073-1076 (2006-03-10)
[reaction: see text] Oxidative C-H amination of N-trichloroethoxysulfonyl-protected ureas and guanidines is demonstrated to proceed in high yield for tertiary and benzylic-derived substrates. The success of these reactions is predicated on the choice of the electron-withdrawn 2,2,2-trichloroethoxysulfonyl (Tces) protecting group

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service