Skip to Content
Merck
All Photos(1)

Key Documents

652725

Sigma-Aldrich

Poly(bis(4-iodophenoxy)phosphazene)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C12H8I2NO2P)n
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

form

solid

refractive index

n20/D 1.710

transition temp

Tg 43.1 °C

General description

Poly [(organo) phosphazene] polymers have an inorganic backbone made up of alternating nitrogen and phosphorous atoms. The inorganic atoms are connected by alternating single and double bonds. Poly(bis(4-iodophenoxy)phosphazene) is a poly[(organo) phosphazene] with iodinated side groups. It may be useful as radio-opaque biomaterials.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Iodine-containing radio-opaque polyphosphazenes.
Hindenlang M D, et al.
Polym. Chem., 1(9), 1467-1474 (2010)
Applicative aspects of poly(organophosphazenes) (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service