513172
1,1-Dichloroethylene
99%
Synonym(s):
Vinylidene chloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH2=CCl2
CAS Number:
Molecular Weight:
96.94
Beilstein:
1733365
EC Number:
MDL number:
Recommended Products
vapor density
3.46 (vs air)
vapor pressure
9.68 psi ( 20 °C)
Assay
99%
autoignition temp.
968 °F
expl. lim.
15.5 %
bp
30-32 °C (lit.)
mp
−122 °C (lit.)
density
1.213 g/mL at 20 °C (lit.)
SMILES string
ClC(Cl)=C
InChI
1S/C2H2Cl2/c1-2(3)4/h1H2
InChI key
LGXVIGDEPROXKC-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Product No.
Description
Pricing
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service