Skip to Content
Merck
All Photos(1)

Key Documents

485624

Sigma-Aldrich

Ethane-1,1,2,2-d4

99 atom % D

Synonym(s):

1,1,2,2-Tetradeuteroethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCD2CD2H
CAS Number:
Molecular Weight:
34.09
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

vapor density

1.05 (vs air)

vapor pressure

37.95 mmHg ( 21.1 °C)

isotopic purity

99 atom % D

form

gas

bp

−88 °C (lit.)

mp

−172 °C (lit.)

mass shift

M+4

SMILES string

[2H]C([2H])C([2H])[2H]

InChI

1S/C2H6/c1-2/h1-2H3/i1D2,2D2

InChI key

OTMSDBZUPAUEDD-LNLMKGTHSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Recommended products

Stainless steel control valves Z146951, Z187224 or either of stainless steel regulators Z512966 and Z512974 is recommended.

Pictograms

FlameGas cylinder

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Gas 1 - Press. Gas Liquefied gas

Storage Class Code

2A - Gases

WGK

WGK 3

Flash Point(F)

-211.0 °F - closed cup

Flash Point(C)

-135.00 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service